Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-EPS15 (Tyr849) Polyclonal Antibody, Invitrogen™

Catalog No. PIPA537564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537564 Supplier Thermo Scientific Supplier No. PA537564

Rabbit Polyclonal Antibody

A suggested positive control for Western blot is JK cells.

Phospho-EPS15 (Tyr849) Polyclonal antibody specifically detects Phospho-EPS15 (Tyr849) in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-EPS15 (Tyr849)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPS15
Gene Accession No. P42566, P42567
Gene Alias 2410112D09Rik; AF1P; AF-1P; AF-1p protein; ALL1 fused gene from chromosome 1; epidermal growth factor pathway substrate 15; epidermal growth factor receptor pathway substrate 15; epidermal growth factor receptor substrate 15; EPS15; MLLT5; protein AF-1p; Protein Eps15; RGD1307641
Gene Symbols EPS15
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of tyrosine 849 (S-A-Y(p)-P-S) derived from Human Ephrin EPS15.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13858, 2060, 313474
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.