Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-ErbB2 (HER-2) (Tyr1139) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577968 Supplier Invitrogen™ Supplier No. PA577968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: A431 (200 ng/mL Trichostatin A treatment for 15 min), A431 (200 ng/mL Trichostatin A for 15 min) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ErbB2 (HER-2) (Tyr1139)
Applications Western Blot
Classification Polyclonal
Concentration 0.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene ERBB2
Gene Accession No. P04626
Gene Alias avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb-b2 avian erythroblastic leukemia viral oncoprotein 2; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian)
Gene Symbols ERBB2
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide surrounding phospho Tyr1139 of human Her2/ErbB2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2064
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.