Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-ErbB3 (Ser686) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105528
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105528 Supplier Invitrogen™ Supplier No. PA5105528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of HER3/ErbB3 only when phosphorylated at Ser686.

ErbB3 (c-erbB-3/HER-3) is a member of the type I family of growth factor receptors and binds to ligands in the heregulin family. ErbB3 is over-expressed in a variety of tumors in the prostate, bladder, breast, stomach, pancreas, and colon. Heregulin and EGF stimulate tyrosine phosphorylation of c-erbB-3 to different extents. The ErbB3 gene is located on the long arm of human chromosome 12 (12q13) and is transcribed into a 6. 2kb mRNA which is translated into a 160/180 kDa glycoprotein. The ErbB3 gene encodes a transmembrane receptor tyrosine kinase due to alternative splicing and forms heterodimers with other EGF receptor family members that have kinase activity. Heterodimerization leads to the activation of pathways that lead to cell proliferation or differentiation. Alternate transcriptional splice variants encoding different isoforms of ErbB3 have been characterized. One ErbB3 isoform lacks the intermembrane region, is secreted outside the cell, and acts to modulate the activity of the membrane-bound form. Additional splice variants have also been reported, but they have not been thoroughly characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ErbB3 (Ser686)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ERBB3
Gene Accession No. P21860, Q61526, Q62799
Gene Alias avian erythroblastosis oncogene B 3; avian erythroblastosis oncogene B 3 receptor; C76256; c-erbB3; c-erbB-3; C-errB-3; erb-b2 receptor tyrosine kinase 3; ERBB3; Erbb-3; Erbb3r; erbB3-S; glial growth factor receptor; HER3; human epidermal growth factor receptor 3; LCCS2; MDA BF 1; MDA-BF-1; MGC88033; nuc-ErbB3; p180 ErbB3; p180-ErbB3; p45 sErbB3; p45-sErbB3; p85 sErbB3; p85-sErbB3; proto-oncogene-like protein c-ErbB-3; receptor tyrosine kinase; receptor tyrosine-protein kinase erbB-3; receptor tyrosine-protein kinase erbB-3; LOW QUALITY PROTEIN: receptor tyrosine-protein kinase erbB-3; tyrosine kinase-type cell surface receptor HER3; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Gene Symbols ERBB3
Host Species Rabbit
Immunogen A synthesized peptide derived from human ERBB3(Accession P21860), corresponding to amino acid residues around phosphorylated Ser686.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13867, 2065, 29496
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.