Learn More
Invitrogen™ Phospho-ERK1/ERK2 (Thr202, Tyr204) Recombinant Rabbit Monoclonal Antibody (ERK12T202Y204-A11), APC
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA536898
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
ERK1 and ERK2 are widely expressed and are involved in the regulation of meiosis, mitosis, and postmitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. When growth factors bind to the receptor tyrosine kinase, Ras interacts with Raf, the serine/threonine protein kinase and activates it as well. Once actived, Raf phosphorylates serine residue in 2 further kinases, MEK1/2, which in turn phosphorylates tyrosine/threonine in extracellular-signal regulated kinase (ERK) 1/2. Upon activation, the ERKs either phosphorylate a number of cytoplasmic targets or migrate to the nucleus, where they phosphorylate and activate a number of transcription factors such as c-Fos and Elk-1.
Specifications
Phospho-ERK1/ERK2 (Thr202, Tyr204) | |
Recombinant Monoclonal | |
APC | |
MAPK1 | |
12559; 9030612K14Rik; AA407128; AU018647; BcDNA:RE08694; C78273; CG12559; CG12559-PA; CG12559-PC; CG12559-PD; CG12559-PE; CG12559-PG; CG12559-PH; CG12559-PI; CG18732; CT34260; CT39192; DERK; D-ERK; DERK-A; Diphospho-ERK; dm-dpERK; Dmel\CG12559; Dmel_CG12559; DmErk; DmERKA; DmERK-A; DmMAPK; dpERK; dp-ERK; dpERK1; dpMAPK; drosophila ERK; Dsor2; E(sina)7; EC2-1; EK2-1; enhancer of seven in absentia 7; ERK; Erk MAP kinase; Erk/Map kinase; ERK/rolled; Erk1; Erk-1; erk1 erk2; Erk1/2; ERK1b; ERK2; ERK-2; ERKA; ERK-A; ERT1; ERT2; Esrk1; extracellular signal-regulated kinase; extracellular signal-regulated kinase (ERK2); extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase A; extracellular signal-regulated kinase-1; extracellular signal-regulated kinase-2; extracellular signal-related kinase 1; Extracellular-regulated kinase A; extracellular-signal-regulated kinase 2; Extracellular-signal-related kinase A; EY2-2; fi06b09; GroupII; HS44KDAP; HUMKER1A; I79_009500; I79_018350; insulin-stimulated MAP2 kinase; l(2)41Ac; l(2R)EMS45-39; M phase MAP kinase; MAP kinase; MAP kinase 1; MAP kinase 2; MAP kinase 3; MAP kinase isoform p42; MAP kinase isoform p44; MapK; MAP-k; MAPK 1; MAPK 2; MAPK 3; MAPK1; mapk1.S; mapk1a; mapk1-a; mapk1-b; mapk2; mapk3; MAP-kinase; MBP kinase; Microtubule-associated protein 2 kinase; mitogen activated protein kinase; mitogen activated protein kinase 1; mitogen activated protein kinase 3; mitogen-activated 3; mitogen-activated protein kinase; mitogen-activated protein kinase 1; mitogen-activated protein kinase 1 S homeolog; mitogen-activated protein kinase 1a; mitogen-activated protein kinase 2; Mitogen-activated protein kinase 3; Mitogen-activated protein kinase ERK-A; MNK1; mpk1; Mtap2k; Myelin basic protein kinase; Myelin xP42 protein kinase; p38; p40; p41; p41mapk; p42; p42 MAP Kinase; P42MAPK; p42-MAPK; p44; p44 MAP kinase; p44erk1; p44-ERK1; p44mapk; p44-MAPK; pERK; p-ERK; phospho-ERK; pMapK; pp42/MAP kinase; Prkm1; PRKM2; Prkm3; protein kinase; Protein rolled; protein tyrosine kinase ERK2; RE08694p; RL; rl/ERK; rl/MAPK; rll; rl-PA; rl-PC; rl-PD; rl-PE; rl-PG; rl-PH; rl-PI; roll; rolled; sem; sevenmaker; SR2-1; Su(Raf)2B; wu:fi06b09; XELAEV_18010534mg; xp42; zERK1; zERK2 | |
Rabbit | |
Protein A/G | |
RUO | |
26413, 26417, 5594, 5595 | |
4°C | |
Liquid |
Flow Cytometry | |
ERK12T202Y204-A11 | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P27361, P28482, P63085, Q63844 | |
MAPK1, MAPK3 | |
A synthetic phospho-peptide corresponding to residues surrounding Thr202/Tyr204 of human phospho Erk1/2. | |
10 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.