Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Phospho-ERK1 (T202/Y204)/ERK2 (T185/Y187) DuoSet IC ELISA
SDP

Catalog No. DYC1018B2 Shop All R&D Systems Products
Change view
Click to view available options
Format:
15 x 96-well Plates
2 x 96-well Plates
5 x 96-well Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
DYC1018B2 2 x 96-well Plates
DYC1018BE 15 x 96-well Plates
DYC1018B5 5 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC1018B2 Supplier R&D Systems™ Supplier No. DYC1018B2
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Phospho-ERK1 (T202/Y204)/ERK2 (T185/Y187) DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Phospho-ERK1 (T202/Y204)/ERK2 (T185/Y187) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat ERK1/ERK2.

Specifications

Product Type ELISA Kit
Quantity 1 Kit (for 2 Plates)
Species Human, Mouse, Rat
Conjugate Biotin
Assay Range 469.00 to 30000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 2 x 96-well Plates
For Use With (Application) ELISA
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.