Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Phospho-ERK1 (T202/Y204)/ERK2 (T185/Y187) DuoSet IC ELISA
Shop All Novus Biologicals Products

Description
The Phospho-ERK1 (T202/Y204)/ERK2 (T185/Y187) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat ERK1/ERK2.
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 1 Kit (for 5 Plates) |
| Species | Human, Mouse, Rat |
| Conjugate | Biotin |
| Assay Range | 469.00 to 30000 pg/mL |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Format | 5 x 96-well Plates |
| For Use With (Application) | ELISA |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?