Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-ETK (Tyr40) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536787
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536787 Supplier Invitrogen™ Supplier No. PA536787
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 78 kDa. Purity is >95% by SDS-PAGE.

Bone marrow tyrosine kinase (BMX) is a non-receptor tyrosine kinase that belongs to the SRC-related TEC subfamily of tyrosine kinases. It is an important regulator of various cellular processes including apoptosis, cell survival, cellular differentiation, cell migration, and transformation. One family of nonreceptor TKs includes the genes TEC (MIM 600583), TXK (MIM 600058), ITK (MIM 186973), and BTK (MIM 300300). All of these proteins are homologs of the Drosophila Src28 TK and contain an SH3 and SH2 domain upstream of the TK domain.[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ETK (Tyr40)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.2
Gene BMX
Gene Accession No. P51813, P97504
Gene Alias BMX; BMX non-receptor tyrosine kinase; bone marrow tyrosine kinase gene in chromosome X protein; Bone marrow tyrosine kinase gene in chromosome X protein homolog; BTK-like on X chromosome; cytoplasmic tyrosine-protein kinase BMX; epa3; Epithelial and endothelial tyrosine kinase; ETK; Etk/Bmx; Etk/Bmx cytosolic tyrosine kinase; NTK38; NTK38 tyrosine kinase; PSCTK2; PSCTK3; RGD1565643; Tyro8; TYRO8 protein tyrosine kinase 8
Gene Symbols BMX
Host Species Rabbit
Immunogen Synthetic phosphopeptide derived from human ETK around the phosphorylation site of Tyrosine 659.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12169, 367786, 660
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.