Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-ETK (Tyr566) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538414
Description
ETK, also known as BMX, is a member of the Bruton′s tyrosine kinase (Btk) family. It is expressed in a variety of hematopoietic, epithelial and endothelial cells. Etk, like other Btk family members, contains a pleckstrin homology (PH) domain and Src homology SH3 and SH2 domains. It participates in multiple signal transduction pathways. Phosphorylation of Tyr566 by Src kinase is required for activation of Etk in vivo. In endothelial and epithelial cells, Etk is regulated by FAK through phosphorylation at Tyr40.
Specifications
| Phospho-ETK (Tyr566) | |
| Polyclonal | |
| Unconjugated | |
| BMX | |
| Cytoplasmic tyrosine-protein kinase BMX, Bone marrow tyrosine kinase gene in chromosome X protein, Epithelial and endothelial tyrosine kinase, NTK38, BMX, ETK | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12169, 660 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51813, P97504 | |
| BMX | |
| A synthetic phosphopeptide derived from human ETK around the phosphorylation site of Tyr566 (D-Q-YP-V-S) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction