Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho Exo1 (Ser746) Rabbit anti-Human,Monkey, KHL, Polyclonal, MilliporeSigma™

Catalog No. ABE106610MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE106610MI 100 μL
ABE106625MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE106610MI Supplier MilliporeSigma Supplier No. ABE1066100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

phospho Exo1 (Ser746) Polyclonal antibody specifically detects phospho Exo1 (Ser746) in Human,Monkey samples. It is validated for Dot Blot, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho Exo1 (Ser746)
Applications Dot Blot, Western Blot
Classification Polyclonal
Conjugate KLH
Gene Accession No. NP_006018
Gene Alias hExo1;Exonuclease 1;hExoI
Gene Symbols EXOi, EXOI, HEX1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphoserine 746 from the C-terminal region of human Exonuclease 1.
Molecular Weight of Antigen ∼110 kDa observed; 94.10 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 100 μL
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Exonuclease 1 phosphorylated on Serine 746. It targets an epitope within 10 amino acids surrounding the phosphoserine 746 residue from the C-terminal region.
Target Species Human, Monkey
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.