Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Ezrin/Radixin/Moesin (Thr558, Thr567, Thr564) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117253
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117253 Supplier Invitrogen™ Supplier No. PA5117253
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: PC-12.

Moesin, a member of the talin-4.1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Ezrin/Radixin/Moesin (Thr558, Thr567, Thr564)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ezr
Gene Accession No. O35763, P15311, P26038, P31977, P35241
Gene Alias AA516625; AW146364; C78546; CVIL; CVL; cytovillin; cytovillin 2; DFNB24; epididymis luminal protein 70; epididymis secretory protein Li 105; ESP10; EZR; Ezrin; HEL70; HEL-S-105; membrane-organizing extension spike protein; Moesin; MSN; p81; R75297; radixin; radixin isoform; RDX; VIL2; villin 2; villin 2 (ezrin); villin-2
Gene Symbols Ezr, MSN, RDX
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315655, 4478, 54319, 5962, 7430, 81521
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.