Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-FANCA (Ser1149) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103268
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103268 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103268 Supplier Invitrogen™ Supplier No. PA5103268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of FANCA only when phosphorylated at Ser1149.

This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that are known calcium sensors and mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. The encoded protein is also a substrate for ubiquitin-E3-ligase parkin. The gene has previously been referred to as synaptotagmin XII but has been renamed synaptotagmin XI to be consistent with mouse and rat official nomenclature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-FANCA (Ser1149)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FANCA
Gene Accession No. O15360, Q9JL70
Gene Alias AW208693; FA; FA1; FAA; FACA; FAH; FA-H; Fanca; FANCH; Fanconi anemia complementation group A; Fanconi anemia group A protein; Fanconi anemia group A protein homolog; Fanconi anemia, complementation group A; Fanconi anemia, complementation group H; Fanconi anemia, type 1; Protein FACA
Gene Symbols FANCA
Host Species Rabbit
Immunogen A synthesized peptide derived from human FANCA(Accession O15360), corresponding to amino acid residues around phosphorylated Ser1149.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14087, 2175, 361435
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.