Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-FANCD2 (Ser717) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106174
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106174 Supplier Invitrogen™ Supplier No. PA5106174
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of FANCD2 only when phosphorylated at Ser717.

FANCD2 (Fanconi anemia group D2 protein) is required for maintenance of chromosomal stability. It promotes accurate and efficient pairing of homologs during meiosis. FANCD2 is involved in the repair of DNA double-stranded breaks, both by homologous recombination and single-stranded annealing. It is required for the targeting/stabilization of BLM to non-centromeric abnormal structures induced by replicative stress. FANCD2 promotes BRCA2/FANCD1 loading onto damaged chromatin. Defects in the FANCD2 gene result in Fanconi anemia complementation group D2. This disorder affects all bone marrow elements resulting in anemia, leukopenia, and thrombopenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-FANCD2 (Ser717)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FANCD2
Gene Accession No. Q80V62, Q9BXW9
Gene Alias 2410150O07Rik; AU015151; BB137857; DKFZp762A223; FA4; FACD; FACD2; FAD; FAD2; FA-D2; FANCD; FANCD2; Fanconi anemia complementation group D2; Fanconi anemia D2 protein; Fanconi anemia group D2 protein; Fanconi anemia group D2 protein homolog; Fanconi anemia, complementation group D2; FLJ23826; Protein FACD2
Gene Symbols FANCD2
Host Species Rabbit
Immunogen A synthesized peptide derived from human FANCD2(Accession Q9BXW9), corresponding to amino acid residues around phosphorylated Ser717.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 211651, 2177
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.