Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-FAS (Tyr291) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538490
Description
Fas ligand (FasL, CD 95L) is a type-II-membrane protein, whose N-terminus is in the cytoplasm and its C-terminal region extends into the extracellular space. Its receptor Fas (CD 95, Apo-1) is a cell-surface-type-I-membrane protein and a member of the tumor necrosis factor (TNF) and nerve growth factor (NGF) receptor family. As a member of the TNF-cytokine family FasL induces apoptosis when interacting with its receptor Fas. FasL may exist as either membrane bound (45 kD) or soluble forms (26 kD). The soluble protein can be released from cells upon cleavage by metalloproteinases. Binding of FasL to Fas leads to oligomerization of the receptor and triggers apoptotic cell death through the interaction of other proteins. FasL is predomitly expressed in activated T-lymphocytes and natural killer (NK) cells also it is expressed in the tissues of immune-privilege sites such as testis and eye. FasL expression is also reported in various tissues as thymus, liver, ovary, lung, heart and kidney. The Fas/FasL system has been shown to play a role in a number of human diseases, for example AIDS, hepatitis or cancer. It is assumed that induction of apoptosis through FasL is predomitly involved in anti-viral immune responses.
Specifications
Phospho-FAS (Tyr291) | |
Polyclonal | |
Unconjugated | |
FAS | |
ALPS1A, APO-1, APT1, Apo-1 Fas, CD95, FAS1, FASTM, TNFRSF6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
355 | |
-20°C | |
Liquid |
Peptide ELISA, Immunofluorescence | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P25445 | |
FAS | |
A synthetic phosphopeptide derived from human FAS around the phosphorylation site of Tyr291 (E-A-YP-D-T) | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction