Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Fascin (Ser39) Monoclonal Antibody (M315)
GREENER_CHOICE

Catalog No. PIMA547627
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547627 Supplier Invitrogen™ Supplier No. MA547627
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody detects a 55 kDa* band corresponding to fascin on SDS-PAGE immunoblots of mouse C2C12 cells treated with Calyculin A. This band is not observed after lambda phosphatase treatment.

Fascin organizes filamentous actin into bundles with a minimum of 4. 1:1 actin/fascin ratio. It is probably involved in the assembly of actin filament bundles present in microspikes, membrane ruffles, and stress fibers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Fascin (Ser39)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone M315
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene FSCN1
Gene Accession No. P85845, Q16658, Q61553
Gene Alias 55 kDa actin-bundling protein; 55kDa actin-bundling protein; AI663989; Fan1; fascin; fascin 1; fascin actin-bundling protein 1; Fascin homolog 1; fascin homolog 1, actin bundling protein; fascin homolog 1, actin bundling protein (Strongylocentrotus purpuratus); fascin homolog 1, actin-bundling protein; Fascin1; fascin-1; Fscn1; fscn1 {ECO:0000250; HSN; p55; Singed; Singed (Drosophila) like (sea urchin fascin homolog like); singed-like (fascin homolog, sea urchin); singed-like protein; SNL; UniProtKB:Q16658}
Gene Symbols FSCN1
Host Species Mouse
Immunogen Clone (M315) was generated from a phospho-Fascin (Ser39) synthetic peptide (coupled to carrier peptide) corresponding to amino acids surrounding Ser39 in human Fascin. This sequence has high homology to the conserved site in rat and mouse Fascin, as well as to the conserved region in Fascin 2, but low homology to the conserved region in Fascin 3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14086, 6624, 683788
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.