Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-FGFR Pan (Tyr653, Tyr654) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564626 Supplier Invitrogen™ Supplier No. PA564626
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-FGFR Pan (Tyr653, Tyr654) Polyclonal Antibody detects endogenous levels of FGFR Pan only when phosphorylated at Tyr653, Tyr654.

This gene encodes a protein that is a member of the fibroblast growth factor receptor family, where amino acid sequence is highly conserved between members and throughout evolution. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein consists of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. This particular family member binds both acidic and basic fibroblast growth factors and is involved in limb induction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-FGFR Pan (Tyr653, Tyr654)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FGFR1
Gene Accession No. P11362, P16092, P21802, P21803, P22455, P22607, Q03142, Q04589, Q498D6, Q61851
Gene Alias ACH; AU043015; AW208770; AW556123; bacteria-expressed kinase; basic fibroblast growth factor receptor 1; BBDS; BEK; BEK fibroblast growth factor receptor; BFGFR; bFGF-R; bFGF-R-1; BFR-1; CD331; CD332; CD333; CD334; CEK; CEK2; CEK3; CFD1; c-fgr; CTLA-2-beta protein; Eask; ECCL; ECT1; FGF receptor-1; FGFBR; fgfr 1; FGFR1; Fgfr-1; FGFR1/PLAG1 fusion; FGFR2; Fgfr-2; FGFR3; FGFR-3; FGFR4; FGFR-4; Fgfr7; Fgfr-7; fibroblast growth factor receptor 1; Fibroblast growth factor receptor 2; fibroblast growth factor receptor 3; fibroblast growth factor receptor 3 variant 4; fibroblast growth factor receptor 4; fibroblast growth factor receptor 4 16 minus form; Flg; Flg-2; FLT2; FLT-2; Fms-like tyrosine kinase 2; fms-related tyrosine kinase 2; FR3; HBGFR; heparin-binding growth factor receptor; HH2; HRTFDS; HSFGFR3EX; Hspy; hydroxyaryl-protein kinase; JTK2; JTK4; JWS; KAL2; Keratinocyte growth factor receptor; KGFR; KGFRTr; KSAM; K-sam; MFR; Mfr3; Mpk-11; N-sam; OGD; protein tyrosine kinase, receptor like 14; protein-tyrosine kinase; protein-tyrosine kinase receptor MPK-11; Proto-oncogene c-Fgr; sam3; svs; TK14; TK25; TKF; tyrosine kinase JTK4; tyrosine kinase related to fibroblast growth factor receptor; tyrosylprotein kinase
Gene Symbols FGFR1, FGFR2, FGFR3, FGFR4
Host Species Rabbit
Immunogen A synthesized peptide derived from human FGFR1/2/3/4 (Phospho-Tyr653+Tyr654).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14182, 14183, 14184, 14186, 2260, 2261, 2263, 2264, 25022, 25114, 79114, 84489
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.