Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118586
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118586 Supplier Thermo Scientific Supplier No. PA5118586
Add to Cart
Add to Cart

Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) Polyclonal Antibody

Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-FGFR1/FGFR2 (Tyr463, Tyr466)
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FGFR1
Gene Accession No. P11362, P16092, P21802, P21803, Q04589
Gene Alias AU043015; AW208770; AW556123; bacteria-expressed kinase; basic fibroblast growth factor receptor 1; BBDS; BEK; BEK fibroblast growth factor receptor; BFGFR; bFGF-R; bFGF-R-1; BFR-1; CD331; CD332; CEK; CEK3; CFD1; c-fgr; Eask; ECCL; ECT1; FGF Receptor 2; FGF receptor-1; FGFBR; fgfr 1; Fgfr1; Fgfr-1; FGFR1/PLAG1 fusion; FGFR2; Fgfr-2; Fgfr7; Fgfr-7; Fibroblast growth factor receptor 1; fibroblast growth factor receptor 2; FLG; FLT2; FLT-2; Fms-like tyrosine kinase 2; fms-related tyrosine kinase 2; HBGFR; heparin-binding growth factor receptor; HH2; HRTFDS; Hspy; hydroxyaryl-protein kinase; JWS; KAL2; Keratinocyte growth factor receptor; KGFR; KGFRTr; KSAM; K-SAM; MFR; N-sam; OGD; protein tyrosine kinase, receptor like 14; proto-oncogene c-Fgr; Soluble KGFR; svs; TK14; TK25
Gene Symbols FGFR1, FGFR2
Host Species Rabbit
Immunogen A synthesized peptide derived from human FGFR1/2 around the phosphorylation site of Tyr463/466.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14182, 14183, 2260, 2263, 25022, 79114
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.