Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-FGFR2 (Ser782) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564796 Supplier Invitrogen™ Supplier No. PA564796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-FGFR2 (Ser782) Polyclonal Antibody detects endogenous levels of FGFR2 only when phosphorylated at Ser782.

FGFR2 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy. FGFR family members differ from one another in their ligand affinities and tissue distribution. Mutations in this gene are associated with Crouzon syndrome, Pfeiffer syndrome, Craniosynostosis, Apert syndrome, Jackson-Weiss syndrome, Beare-Stevenson cutis gyrata syndrome, Saethre-Chotzen syndrome, and syndromic craniosynostosis. Multiple alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-FGFR2 (Ser782)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FGFR2
Gene Accession No. P21802, P21803
Gene Alias AU043015; AW556123; bacteria-expressed kinase; BBDS; BEK; BEK fibroblast growth factor receptor; BFR-1; CD332; CEK3; CFD1; ECT1; FGF Receptor; FGF Receptor 2; FGF-10; FGFR; FGFR2; FGFR-2; Fgfr7; Fgfr-7; fibroblast growth factor receptor 2; JWS; Keratinocyte growth factor receptor; KGFR; KGFRTr; KSAM; K-SAM; protein tyrosine kinase, receptor like 14; Soluble KGFR; svs; TK14; TK25
Gene Symbols FGFR2
Host Species Rabbit
Immunogen A synthesized peptide derived from human FGFR2 (Phospho-Ser782).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14183, 2263
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.