Learn More
Invitrogen™ Phospho-FGFR3 (Tyr724) Polyclonal Antibody

Description
Phospho-FGFR3 (Tyr724) Polyclonal Antibody detects endogenous levels of FGFR3 only when phosphorylated at Tyr724.
Specifications
Specifications
| Antigen | Phospho-FGFR3 (Tyr724) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | FGFR3 |
| Gene Accession No. | P22607, Q61851 |
| Gene Alias | ACH; CD333; CEK2; FGF Receptor; FGFR; FGFR3; FGFR-3; FGFR3deltaTM; Fibroblast growth factor receptor 3; fibroblast growth factor receptor 3 variant 4; Flg-2; FR3; HBGFR; heparin-binding growth factor receptor; HSFGFR3EX; hydroxyaryl-protein kinase; JTK4; Mfr3; sam3; tyrosine kinase JTK4 |
| Gene Symbols | FGFR3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.