Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-FGR (Tyr412) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564583
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564583 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564583 Supplier Invitrogen™ Supplier No. PA564583
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-FGR (Tyr412) Polyclonal Antibody detects endogenous levels of FGR only when phosphorylated at Tyr412.

FGR is a SRC-related kinase present in hematopoietic cells. FGR signals within multiple pathways including within the integrin pathway and downstream of chemokine and cytokine receptors. This protein contains N-terminal sites for myristylation and palmitoylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. It localizes to plasma membrane ruffles, and functions as a negative regulator of cell migration and adhesion triggered by the beta-2 integrin signal transduction pathway. Infection with Epstein-Barr virus results in the overexpression of this protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-FGR (Tyr412)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FGR
Gene Accession No. P09769
Gene Alias c-fgr; c-fgr protooncogene; c-src2; c-src-2 proto-oncogene; EC 2.7.10.2; feline Gardner-Rasheed sarcoma viral oncogene; feline Gardner-Rasheed sarcoma viral oncogene homolog; FGR; FGR protein; FGR proto-oncogene, Src family tyrosine kinase; FLJ43153; Gardner-Rasheed feline sarcoma viral (Fgr) oncogene homolog; Gardner-Rasheed feline sarcoma viral (v-fgr) oncogene homolog; MGC75096; OTTHUMP00000003603; OTTHUMP00000003604; p55c-fgr; p55-c-fgr protein; p55-Fgr; p58c-Fgr; p58-Fgr; proto-oncogene c-Fgr; proto-oncogene tyrosine-protein kinase FGR; SRC2; Tyrosine-protein kinase Fgr; v-fgr feline Gardner-Rasheed sarcoma viral oncogene homolog
Gene Symbols FGR
Host Species Rabbit
Immunogen A synthesized peptide derived from human FGR (Phospho-Tyr412).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2268
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.