Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-FHIT (Tyr114) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985175
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5175 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5175 Supplier Bioss Supplier No. BS3146R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Alterations and deletions of the FHIT gene are strongly linked to the genesis and establishment of human tumors of the lung, cervix, breast, colon, stomach and pancreas. In normal cells, FHIT may act as a tumor suppressor.

Specifications

Antigen Phospho-FHIT (Tyr114)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FHIT
Gene Accession No. P49789
Gene Alias AP3A hydrolase; AP3Aase; AW045638; Bis(5′-adenosyl)-triphosphatase; bis5′-adenosyl-triphosphatase; Diadenosine 5′,5′′′-P1,P3-triphosphate hydrolase; Dinucleosidetriphosphatase; Fhit; Fra14A2; FRA3B; fragile histidine triad; fragile histidine triad gene; Fragile histidine triad protein
Gene Symbols FHIT
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human FHIT around the phosphorylation site of Tyr114.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2272
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.