Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-FHOD1 (Thr1141) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143701 Supplier Invitrogen™ Supplier No. PA5143701
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody was cross-adsorbed to unphosphorylated FHOD1 (Thr-1141) peptide before affinity purification using phospho-FHOD1 (Thr-1141) peptide (without carrier). The antibody detects a 140 kDa* protein corresponding to the apparent molecular mass of FHOD1 on SDS-PAGE immunoblots of human K562 and mouse C2C12 cells treated with calyculin A.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-FHOD1 (Thr1141)
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene FHOD1
Gene Accession No. Q6P9Q4, Q9Y613
Gene Alias BC017144; FH1/FH2 domain-containing protein 1; Fhod1; FHOS; FHOS1; formin homolog overexpressed in spleen 1; formin homology 2 domain containing 1; formin homology 2 domain-containing protein 1
Gene Symbols FHOD1
Host Species Rabbit
Immunogen Phospho-FHOD1 (Thr-1141) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding Thr-1141 in human FHOD1. This peptide sequence is highly conserved in rat and mouse FHOD1, and is well conserved in FHOD3 (Thr-1399).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234686, 29109, 291964
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.