Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-FLNC 2233 (Ser2233) Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13183R

Description
This gene encodes one of three related filamin genes, specifically gamma filamin. These filamin proteins crosslink actin filaments into orthogonal networks in cortical cytoplasm and participate in the anchoring of membrane proteins for the actin cytoskeleton. Three functional domains exist in filamin: an N-terminal filamentous actin-binding domain, a C-terminal self-association domain, and a membrane glycoprotein-binding domain. Two transcript variants encoding different isoforms have been found for this gene.Specifications
Phospho-FLNC 2233 (Ser2233) | |
Polyclonal | |
Unconjugated | |
FLNC | |
1110055E19Rik; ABP-280; ABP280A; ABP-280-like protein; ABPA; Abpl; ABP-L; ABP-L, gamma filamin; actin binding protein 280; actin-binding protein 280; Actin-binding-like protein; filamin C; Filamin C gamma; filamin C, gamma; filamin C, gamma (actin binding protein 280); filamin, gamma; Filamin-2; Filamin-C; FLJ10186; Fln2; FLNC; FLN-C; gamma filamin; Gamma-filamin; MFM5; MPD4 | |
Rabbit | |
Protein A | |
RUO | |
2318 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q14315 | |
FLNC | |
KLH conjugated synthetic phosphopeptide derived from human FLNC around the phosphorylation site of Ser2233. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction