Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-FOXO3A (Ser315) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564553
Description
Phospho-FOXO3 (Ser315) Polyclonal Antibody detects endogenous levels of FOXO3 only when phosphorylated at Ser315.
USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
Specifications
Phospho-FOXO3A (Ser315) | |
Polyclonal | |
Unconjugated | |
FOXO3 | |
1110048B16Rik; 2010203A17Rik; AF6q21; AF6q21 protein; C76856; Fkhr2; FKHRL1; FKHRL1P2; forkhead box O3; forkhead box O3a; forkhead box O3A transcription factor; forkhead box protein O3; forkhead homolog (rhabdomyosarcoma) like 1; forkhead in rhabdomyosarcoma-like 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; FOXO2; FOXO3; FOXO-3; FOXO3A | |
Rabbit | |
Affinity chromatography | |
RUO | |
2309, 294515, 56484 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide; pH 7.3 | |
O43524, Q9WVH4 | |
FOXO3 | |
A synthesized peptide derived from human FKHRL1 (Phospho-Ser315). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction