Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-FoxO3a (Ser318, Ser321) Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3138R

Description
USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.Specifications
Phospho-FoxO3a (Ser318, Ser321) | |
Polyclonal | |
Unconjugated | |
FOXO3 | |
1110048B16Rik; 2010203A17Rik; AF6q21; AF6q21 antibody; AF6q21 protein; AFX; AFX1; Afxh; BOS_9647; C76856; DAF16 antibody; DKFZp781A0677; FkhR; Fkhr2; FKHR2 antibody; FKHRL1; FKHRL1 antibody; FKHRL1P2; FKHRL1P2 antibody; FKHR-LIKE 1; Forkhead Box O1; forkhead box O3; forkhead box O3A; forkhead box O3A transcription factor; Forkhead Box O4; Forkhead box protein O3; forkhead homolog (rhabdomyosarcoma) like 1; Forkhead in rhabdomyosarcoma-like 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; FOXO1; FOXO2; Foxo3; FOXO-3; FOXO3A; FOXO4; in rhab; MGC120490; MGC12739; MGC31925; MLLT7 | |
Rabbit | |
Protein A | |
RUO | |
2309, 294515, 56484 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O43524, Q9WVH4 | |
FOXO3 | |
KLH conjugated synthetic phosphopeptide derived from human FoxO3a around the phosphorylation site of Ser318/321. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction