Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-FOXO4/FOXO1 (Ser197, Ser256) Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3144R

Description
These genes encode a member of the O class of winged helix/forkhead transcription factor family. Proteins encoded by this class are regulated by factors involved in growth and differentiation indicating they play a role in these processes. Multiple transcript variants encoding different isoforms have been found for these genes. Both function as transcription factors involved in the regulation of the insulin signaling pathway.Specifications
Phospho-FOXO4/FOXO1 (Ser197, Ser256) | |
Polyclonal | |
Unconjugated | |
FOXO4 | |
FOXO1, FOXO4 | |
KLH conjugated synthetic phosphopeptide derived from human FOXO4 around the phosphorylation site of Ser197. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P98177, Q12778 | |
Rabbit | |
Protein A | |
RUO | |
2308, 4303 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction