Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-FSCN1 (Ser39) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980499
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0499 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0499 Supplier Bioss Supplier No. BS0772R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fascin organizes filamentous actin into bundles with a minimum of 4. 1:1 actin/fascin ratio. It is probably involved in the assembly of actin filament bundles present in microspikes, membrane ruffles, and stress fibers.

Specifications

Antigen Phospho-FSCN1 (Ser39)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FSCN1
Gene Accession No. Q16658
Gene Alias 55 kDa actin-bundling protein; AI663989; Fan1; Fascin; fascin 1; fascin actin-bundling protein 1; fascin homolog 1, actin bundling protein; fascin homolog 1, actin bundling protein (Strongylocentrotus purpuratus); fascin homolog 1, actin-bundling protein; fascin-1; Fscn1; fscn1 {ECO:0000250; HSN; p55; Singed; singed-like (fascin homolog, sea urchin); Singed-like protein; SNL; UniProtKB:Q16658}
Gene Symbols FSCN1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human FSCN1 around the phosphorylation site of Ser39.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6624
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.