Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-FXYD1 (Ser83) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986505
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6505 Supplier Bioss Supplier No. BS5348R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the FXYD family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The protein encoded by this gene is a plasma membrane substrate for several kinases, including protein kinase A, protein kinase C, NIMA kinase, and myotonic dystrophy kinase. It is thought to form an ion channel or regulate ion channel activity and act as an accessory protein of Na,K-ATPase. Alternatively spliced transcript variants have been described.

Specifications

Antigen Phospho-FXYD1 (Ser83)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FXYD1
Gene Accession No. O00168
Gene Alias 0610012C17Rik; 1110006M24Rik; FXYD domain containing ion transport regulator 1; FXYD domain-containing ion transport regulator 1; Fxyd1; Phospholemman; PLM; Pml; Sodium Potassium ATPase; Sodium/potassium-transporting ATPase subunit FXYD1
Gene Symbols FXYD1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human FXYD1 around the phosphorylation site of Ser83.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5348
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.