Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-GAB1 (Tyr627) Polyclonal Antibody

Catalog No. 44568G
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-568-G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-568-G Supplier Invitrogen™ Supplier No. 44568G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Phospho-GAB1 (Tyr627) Polyclonal Antibody for Western Blot

Gab1 is a 115 kDa multiple docking protein that plays an essential role in cellular growth, transformation and apoptosis. Gab1 can be phosphorylated by multiple receptor tyrosine kinase (RTKs), including: insulin receptor (IR), platelet-derived growth factor receptor beta] (PDGFRbeta]), hepatocyte growth factor/scatter factor receptor (HGFR/SFR or c-Met), and epidermal growth factor receptor (EGF), as well as in response to cell-cell adhesion. Gab1 is tyrosine phosphorylated on at least 16 sites, some of which serve as binding sites for phosphoatidylinositol 3-kinase (PI3K), Grb2, PLC gamma 1, Nck, and SHP2. Phosphorylation of Gab1 on tyrosines 627 and 659 is critical for its binding to SHP2, and for activation of the ERK/MAPK pathway in response to EGF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GAB1 (Tyr627)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
Gene GAB1
Gene Accession No. Q13480, Q9QYY0
Gene Alias AA408973; AW107238; EGK_16125; GAB1; GRB2 associated binding protein 1; GRB2-associated binder 1; GRB2-associated binding protein 1; GRB2-associated binding protein 1 long isoform; GRB2-associated-binding protein 1; Growth factor receptor bound protein 2-associated protein 1; OTTHUMP00000220263
Gene Symbols GAB1
Host Species Rabbit
Immunogen The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human Gab1 that contains tyrosine 627. The sequence is conserved in mouse.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14388, 2549
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.