Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-GAB1 (Tyr659) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538415
Description
Gab 1 (GRB2-associated binder 1), one such member of the IRS family, plays an important role in cellular growth response, transformation and apoptosis. Gab 1 is a multi-substrate docking protein that functions downstream in the signaling pathways of different receptor kinases, including EGFR. Gab1 is tyrosine phosphorylated normally in response to insulin and consequently enhances phosphatidylinositol 3- kinase (PI3K) binding. In response to osmotic shock, tyrosine-phosphorylated Gab 1 (p-Gab 1) also binds and activates phosphatidylinositol 3-kinase, suggesting that Gab 1 is the major site for PI3K recruitment following osmotic shock stimulation.
Specifications
Phospho-GAB1 (Tyr659) | |
Polyclonal | |
Unconjugated | |
GAB1 | |
GRB2-associated-binding protein 1, GRB2-associated binder 1, Growth factor receptor bound protein 2-associated protein 1, GAB1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14388, 2549, 361388 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q13480, Q9QYY0 | |
GAB1 | |
A synthetic phosphopeptide derived from human GAB1 around the phosphorylation site of Tyr659 (V-D-YP-V-V) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction