Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-GAB2 (Ser159) Polyclonal Antibody
Phospho-GAB2 (Ser159) Polyclonal Antibody
Supplier: Thermo Scientific PA5105241
Description
Phospho-GAB2 (Ser159) Polyclonal Antibody for Western Blot

Specifications
Phospho-GAB2 (Ser159) | |
Polyclonal | |
Unconjugated | |
GAB2 | |
AI463667; D130058I17Rik; GAB2; Grb2 associated binder 2; GRB2 associated binding protein 2; GRB2-associated binder 2; GRB2-associated binding protein 2; GRB2-associated-binding protein 2; GRB3-3; Growth factor receptor bound protein 2-associated protein 2; KIAA0571; p97; PH domain-containing adaptor molecule p97; pp100 | |
Rabbit | |
Sequential Chromatography | |
RUO | |
14389, 84477, 9846 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9EQH1, Q9UQC2, Q9Z1S8 | |
GAB2 | |
A synthesized peptide derived from human GAB2(Accession Q9UQC2), corresponding to amino acid residues around phosphorylated Ser159. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction