Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ phospho-GADS (Thr262), Mouse, Unlabeled, Clone: 6E11.2,

Catalog No. MABS1187MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1187MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1187MI Supplier MilliporeSigma™ Supplier No. MABS1187
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GRB2-related adapter protein 2 (UniProt O75791; also known as Adapter protein GRID, GRAP-2, GRB-2-like protein, Grb2-related adapter protein downstream of Shc, GRB2L, GRBLG, GRBX, Grf-40, Grf40 adapter protein, Growth factor receptor-binding protein, Hematopoietic cell-associated adapter protein GrpL, P38, Protein GADS, SH3-SH2-SH3 adapter Mona) is encoded by the GRAP2 (also known as GADS, GRB2L, GRID) gene (Gene ID 9402) in human. GADS (Grb2-related adapter protein downstream of Shc), GRB2 and GRAP constitute the GRB2 family of adapter proteins characterized by a central SH2 domain (a.a. 58-149 of GADS) flanked between two SH3 domains (a.a. 1-56, 271-330 of GADS). Although the SH2 domains of all three members have similar binding specificities, the C-terminal SH3 domain of GADS possesses unique binding specificity that allows for its interaction with the adapter protein SLP-76 (SH2 domain-containing leukocyte protein of 76 kDa). The GADS-SLP-76 complex is recruited to tyrosine-phosphorylated LAT (linker for activation of T cells) via GADS SH2 domain, allowing the assembly of signaling complexes critical to T-cell activation. In addition, evidences indicate that GADS plays a critical role in the development of BCR-ABL-mediated lymphoid disease. The oncogenic Bcr-Abl fusion protein is shown to bind GADS in a phosphotyrosine-dependent manner and GADS is expressed in a subset of CML cell lines and B-ALL patient samples, where it associates with both BCR-ABL and SLP-76.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-GADS (Thr262)
Applications Western Blot
Classification Monoclonal
Clone 6E11.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene GRAP2, GADS, GRB2L, GRID
Gene Accession No. O75791
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to a human GADS sequence containing phosphorylated Thr262.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001278753
Test Specificity Clone 6E11.2 detects GADS phosphorylated on Thr262. The corresponding phosphorylation site in human GADS spliced isoform 2 is Thr149 (UniProt O75791-2).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.