Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-GAP43 (Ser41) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14626 Supplier Invitrogen™ Supplier No. PA14626
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, canine, chicken, finch, human, mouse, non-human primate, Xenopus and zebrafish based on 100% sequence homology. This antibody is specific for the ~50 kDa Gap-43 protein phosphorylated at Ser41 in Western blots of rat brain lysate. In some tissues the antibody also recognizes a higher molecular weight protein, that is also recognized by the Pan Gap-43 antibody, that may be a Gap-43 aggregate or oligomer. Immunolabeling blocked by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide.

GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile 'growth cones' that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GAP43 (Ser41)
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene GAP43
Gene Accession No. P06837, P07936
Gene Alias Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1
Gene Symbols GAP43
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser41 of rat GAP43 conjugated to KLH.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14432, 29423
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.