Learn More
Phospho-GATA1 (Ser142) Polyclonal Antibody, Abbomax
Description
Positive control: K562 Cellular location: Nucleus.
Specifications
Specifications
| Antigen | Phospho-GATA1 (Ser142) |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 25mM tris with proprietary stabilizer and 0.01% sodium azide |
| Gene | Gata1 |
| Gene Accession No. | P15976, P17679 |
| Gene Alias | eryf1; Erythroid transcription factor; erythroid transcription factor 1; GATA binding protein 1; GATA binding protein 1 (globin transcription factor 1); GATA1; GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA-binding protein 1 (globin transcription factor 1); GF1; Gf-1; globin transcription factor 1; HGNC:4170; NFE1; NF-E1; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1; XLANP; XLTDA; XLTT |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.