Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho GATA6 (Ser37) Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABE2897100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2897-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE2897-100 Supplier MilliporeSigma Supplier No. ABE2897100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho GATA6 (Ser37), Cat. No. ABE2897, is a highly specific rabbit polyclonal antibody that targets Transcription factor GATA-6 (GATA-binding factor 6) and has been tested for use in Dot Blot, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho GATA6 (Ser37)
Applications Dot Blot, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias GATA-binding factor 6
Gene Symbols GATA6
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphoserine 37 of human Transcription factor GATA-6.
Molecular Weight of Antigen ∽75 kDa observed; 60.03 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_005248
Test Specificity This rabbit polyclonal antibody detects GATA6 phosphorylated on Serine 37.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.