Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-GCN2 (Thr899) Recombinant Rabbit Monoclonal Antibody (6Q8C6)
GREENER_CHOICE

Catalog No. FEMA551544
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551544 Supplier Invitrogen™ Supplier No. MA551544
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: HLTGM.

EIF2AK4 (GCN2) is a metabolic-stress sensing protein kinase that phosphorylates the alpha subunit of eukaryotic translation initiation factor-2 in response to low amino acid availability. It plays a role as an activator of the integrated stress response required for adapatation to amino acid starvation. Mutations in the gene lead to pulmonary vennocclusive disease 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GCN2 (Thr899)
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 6Q8C6
Concentration 0.253 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene EIF2AK4
Gene Accession No. Q9P2K8
Gene Alias 2610011M03; EIF2AK4; eIF-2-alpha kinase GCN2; eukaryotic translation initiation factor 2 alpha kinase 4; eukaryotic translation initiation factor 2-alpha kinase 4; Gcn2; GCN2 eIF2alpha kinase; GCN2-like protein; general control nonderepressible 2; Kiaa1338; mGCN2; PVOD2
Gene Symbols EIF2AK4
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440275
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.