Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-GEF H1 (Ser886) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984575
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4575 Supplier Bioss Supplier No. BS2201R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GEF-H1 activates Rho GTPases by promoting the exchange of GDP and GTP. Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. GEF-H1 may be involved in epithelial barrier permeability, cell motility and polarization, dendritic spine morphology, antigen presentation, leukemic cell differentiation, cell cycle regulation, innate immune response, and cancer. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Antigen Phospho-GEF H1 (Ser886)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ARHGEF2
Gene Accession No. Q92974
Gene Alias AA408978; Arhgef2; CG10188; CG10188 gene product from transcript CG10188-RB; CG10188-PA; CG10188-PB; CG10188-PC; DKFZp547L106; DKFZp547P1516; Dmel\CG10188; Dmel_CG10188; FLJ45440; GEF; GEFH1; GEF-H1; Guanine nucleotide exchange factor H1; KIAA0651; Lbcl1; LBC′S first cousin; Lfc; LFP40; Lymphoid blast crisis-like 1; Microtubule-regulated Rho-GEF; mKIAA0651; Oncogene LFC; OTTHUMP00000018855; P40; Proliferating cell nucleolar antigen p40; Rho guanine nucleotide exchange factor 2; rho guanine nucleotide exchange factor 2-like protein; Rho/Rac guanine nucleotide exchange factor (GEF) 2; Rho/Rac guanine nucleotide exchange factor 2; Rhobin; uncharacterized protein
Gene Symbols ARHGEF2
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human Arhgef2 around the phosphorylation site of Ser886.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9181
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.