Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Girdin (Ser1416) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143702
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143702 Supplier Invitrogen™ Supplier No. PA5143702
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was cross-adsorbed to unphosphorylated Girdin (Ser-1416) peptide before affinity purification using phospho-Girdin (Ser-1416) peptide. This antibody detects a 250 kDa* protein on SDS-PAGE immunoblots of human A431 cells stimulated with EGF or calyculin A. These reactivities are not observed after lambda phosphatase treatment to dephosphorylate Girdin.

Girdin is a Galpha-interacting protein that can enhance the activation of the protein kinase Akt, remodel the actin cytoskeleton, and is thought to be involved in the regulation of cell migration and cancer metastasis. It has recently been shown that Girdin interacts with Disrupted-in-Schizophrenia 1 (DISC1), a susceptibility gene for major psychiatric disorders. DISC1 is thought to be involved in the migration, positioning and differentiation of dentate granule cells (DGCs) during development; depletion of Girdin or blocking the Girdin-DISC1 interaction results in defects in axonal sprouting in the CA3 region of the hippocampus and overextended migration of mispositioning of DGCs, suggesting the Girdin plays a role in postnatal neurogenesis in the dentate gyrus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Girdin (Ser1416)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene CCDC88A
Gene Accession No. Q3V6T2, Q5SNZ0
Gene Alias A430106J12Rik; AI848406; Akt phosphorylation enhancer; AKT-phosphorylation enhancer; Ape; C130096N06Rik; C330012F17Rik; Ccdc88a; coiled coil domain containing 88A; coiled-coil domain containing 88A; coiled-coil domain-containing protein 88A; D130005J21Rik; G alpha-interacting vesicle-associated protein; Galpha-interacting vesicle-associated protein; girders of actin filament; girders of actin filaments; girdin; GIV; Grdn; HkRP1; Hook-related protein 1; KIAA1212; RGD1306694
Gene Symbols CCDC88A
Host Species Rabbit
Immunogen Phospho-Girdin (Ser-1416) synthetic peptide (coupled to carrier) corresponding to amino acid residues around serine 1416 in human Girdin. This sequence is identical in rat and mouse Girdin, and has low homology to similar regions in other Girdin family members.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108686, 305605, 55704
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.