Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-GIT2 (Tyr592) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105211 Supplier Invitrogen™ Supplier No. PA5105211
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of GIT2 only when phosphorylated at Tyr592.

GTPase-activating protein for the ADP ribosylation factor family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GIT2 (Tyr592)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GIT2
Gene Accession No. Q14161
Gene Alias 1500036H07Rik; 5830420E16Rik; 6430510B20Rik; 9630056M03Rik; ARF GAP GIT2; ARF GTPase activating protein 2; ARF GTPase-activating protein GIT2; B230104M05Rik; CAT2; CAT-2; cool associated tyrosine phosphorylated-2; cool-associated, tyrosine phosphorylated protein 2; cool-interacting tyrosine-phosphorylated protein 2; G protein-coupled receptor kinase interacting ArfGAP 2; G protein-coupled receptor kinase-interactor 2; GIT ArfGAP 2; Git2; GRK-interacting protein 2; KIAA0148; mKIAA0148; Paxillin kinase linker; PKL
Gene Symbols GIT2
Host Species Rabbit
Immunogen A synthesized peptide derived from human GIT2(Accession Q14161), corresponding to amino acid residues around phosphorylated Tyr592.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9815
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.