Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-GKAP1 (Ser25) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105217
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105217 Supplier Invitrogen™ Supplier No. PA5105217
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of GKAP only when phosphorylated at Ser25.

The approximately 142 kDa Tankyrase protein is ubiquitously expressed in mammalian cells and is localized to human telomeres. It has homology to ankyrins and to the catalytic domain of poly (adenosine diphosphate-ribose) polymerase (PARP). Tankyrase binds to TRF1 (telomeric repeat binding factor-1), which negatively regulates the maintenance of telomere length.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GKAP1 (Ser25)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gkap1
Gene Accession No. Q5VSY0, Q5XIG5, Q9JMB0
Gene Alias 42 kD cGMP-dependent protein kinase anchoring protein; 42kDa; 4933400B15Rik; cGMP-dependent protein kinase anchoring protein; cGMP-dependent protein kinase anchoring protein 42kDa; cGMP-dependent protein kinase-anchoring protein of 42 kDa; D13Ertd340e; FKSG21; G kinase anchoring protein 1; G kinase-anchoring protein 1; GKAP1; Gkap42; protein kinase anchoring protein GKAP42
Gene Symbols Gkap1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GKAP1(Accession Q5VSY0), corresponding to amino acid residues around phosphorylated Ser25.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361202, 56278, 80318
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.