Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-GluR2 (Ser880) Rabbit anti-Rat,Human,Mouse,Monkey,Chimpanzee, KHL, Polyclonal, MilliporeSigma™

Catalog No. 07294I25UMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07294I25UMI 25 μg
07294I100MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07294I25UMI Supplier MilliporeSigma Supplier No. 07294I25UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

phospho-GluR2 (Ser880) Polyclonal antibody specifically detects phospho-GluR2 (Ser880) in Rat,Human,Mouse,Monkey,Chimpanzee samples. It is validated for Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-GluR2 (Ser880)
Applications Western Blot
Classification Polyclonal
Conjugate KLH
Gene Accession No. NP_058957
Gene Alias Glutamate receptor 2;Glu-R2;AMPA-selective glutamate receptor 2;GluR-B;GluR-K2;Glutamate receptor ionotropic;AMPA 2;GluA2
Gene Symbols FRIA2, GLUR2
Host Species Rabbit
Immunogen KLH-conjugated Linear Peptide corresponding to 9 amino acids surrounding phosphoserine 880 from the the C-terminal region of Rat Glutamate receptor 2.
Molecular Weight of Antigen ∼110 kDa observed; 98.69 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Anti-phospho-GluR2 (Ser880) is a rabbit polyclonal antibody that detects glutamate receptor 2 phosphorylated on serine 880.
Target Species Rat, Human, Mouse, Monkey, Chimpanzee
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.