Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Glycogen synthase 1 (Ser641) Rabbit anti-Human, Clone: 10C1, Bioss
SDP

Catalog No. 501989613
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9613 Supplier Bioss Supplier No. BSM52159R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Interacts with zinc finger protein A20/TNFAIP3 and inhibits TNF-induced NF-kappa-B-dependent gene expression by interfering with an RIP-or TRAF2-mediated transactivation signal. Increases cell surface CD4(T4) antigen expression. Interacts with HIV-1 matrix protein and is packaged into virions and overexpression can inhibit viral replication. May regulate matrix nuclear localization, both nuclear import of PIC (Preintegration complex) and export of GAG polyprotein and viral genomic RNA during virion production.

Specifications

Antigen Phospho-Glycogen synthase 1 (Ser641)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 10C1
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene Gys1
Gene Accession No. P13807
Gene Alias EC 2.4.1.11; Glycogen [starch] synthase, muscle; glycogen synthase; glycogen synthase 1; glycogen synthase 1 (muscle); glycogen synthase 1, muscle; glycogen synthase 3, brain; GSY; Gys; GYS1; Gys3; MGS
Gene Symbols Gys1
Host Species Rabbit
Immunogen Synthetic peptide derived from human Glycogen synthase 1 (S641), around 630-680aa (phospho S641).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2997
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.