Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-GRASP65 (Thr220/Thr224) Mouse anti-Rat, Clone: LX108, MilliporeSigma™

Catalog No. MABS1282MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1282MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1282MI Supplier MilliporeSigma Supplier No. MABS1282
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect GRASP65 using this mouse monoclonal Anti-phospho-GRASP65 (Thr220/Thr224) Antibody, clone LX108. It is used for Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-GRASP65 (Thr220/Thr224)
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone LX108
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Gorasp1,Grasp65
Host Species Mouse
Immunogen His-tagged recombinant full-length wild type GRASP65 protein treated with mitotic cytosol.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone LX108 specifically recognizes GRASP65 phosphorylated on Thr220/Thr224.
Target Species Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.