Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-GRB10 (Tyr67) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985186
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5186 Supplier Bioss Supplier No. BS3157R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRB7, a SH2 domain protein, has a single SH2 domain at its C-terminal, a central region with similarity to Ras GAP, and a proline-rich N terminus. A related SH2 domain-containing protein, GRB10, exhibits a high degree of homology with GRB7. GRB10 undergoes serine but not tyrosine phosphorylation in response to EGF treatment, but appears to bind to the EGF receptor poorly. GRB10 maps to mouse chromosome 11, in close proximity to the EGF receptor. Similarly, GRB7 maps to the same mouse chromosome near the EGF receptor-related protein HER2.

Specifications

Antigen Phospho-GRB10 (Tyr67)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GRB10
Gene Accession No. Q13322
Gene Alias 5730571D09Rik; AI325020; Grb10; Grb-10; GRB10 adapter protein; GRB10 adaptor protein; GRBIR; GRB-IR; growth factor receptor bound protein 10; Growth factor receptor-bound protein 10; Insulin receptor-binding protein Grb-IR; IRBP; KIAA0207; maternally expressed gene 1; Maternally expressed gene 1 protein; Meg1; mKIAA0207; OTTHUMP00000197988; OTTHUMP00000197989; OTTHUMP00000197990; OTTHUMP00000197991; OTTHUMP00000197993; OTTHUMP00000197994; RGD1566234; RSS
Gene Symbols GRB10
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human GRB10 around the phosphorylation site of Tyr67 [SL (p-Y)SA].
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2887
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.