Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-GRK1 (Ser21) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105659 Supplier Invitrogen™ Supplier No. PA5105659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of GRK1 only when phosphorylated at Ser21.

GRK1 is a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinases. GRK1 mediates rapid desensitization of rod photoreceptors to light by catalyzing phosphorylation of the visual pigment rhodopsin which leads to its deactivation. Defects in GRK1 are known to cause Oguchi disease 2 (also known as stationary night blindness Oguchi type-2). GRK1 can be phosphorylated by PKA and this reduces the ability of GRK1 to phosphorylate rhodopsin in vitro. It is speculated that phosphorylation of GRK1 by PKA in vivo occurs in the dark when cAMP levels in photoreceptor cells are elevated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GRK1 (Ser21)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GRK1
Gene Accession No. Q15835, Q63651, Q9WVL4
Gene Alias G protein-coupled receptor kinase 1; G protein-coupled receptpr kinase 1; GPRK1; G-protein coupled receptor kinase 1; G-protein receptor kinase 1; GRK 1a/b; GRK1; GRK1a; rhodopsin kinase; rhodopsin kinase GRK1; Rhok; RK
Gene Symbols GRK1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GRK1(Accession Q15835), corresponding to amino acid residues around phosphorylated Ser21.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24013, 6011, 81760
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.