Learn More
Phospho-GSK3B (Ser9) Mouse anti-Human, Unconjugated, Clone: 1C9E2, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 675581IG100UL
Description
The Phospho-GSK3B (Ser9) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-GSK3B (Ser9). This antibody recognizes human, mouse, rat antigen. The Phospho-GSK3B (Ser9) antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Phospho-GSK3B (Ser9) | |
Monoclonal | |
2240 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence | |
1C9E2 | |
Unconjugated | |
100 μL | |
2932 | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.