Learn More
Invitrogen™ Phospho-GSK3B (Ser9) Recombinant Rabbit Monoclonal Antibody (RM453)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA562158

Description
This antibody reacts to human GSK-3beta only when phosphorylated at Ser9. There is no cross-reactivity to GSK-3beta that is not phosphorylated. This antibody may also react to mouse or rat Phospho- GSK-3beta (Ser9) as predicted by immunogen homology.
GSK3B is a proline-directed serine/threonine protein kinase involved in energy metabolism, neuronal cell development and body pattern formation. It is a negative regulator of glucose homeostasis and is involved in energy metabolism, inflammation, ER-stress, mitochondrial dysfunction, and apoptotic pathways. GSK3B is a critical component of the Wnt/beta-catenin signaling pathway, and its activity results in the inhibition of Wnt signaling by mediating the degradation of beta-catenin. GSK3B is also involved in the Hedgehog, Notch, and insulin signaling pathways.
Specifications
Phospho-GSK3B (Ser9) | |
Recombinant Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
P49841 | |
GSK3B | |
A phospho-peptide corresponding to human phospho-GSK-3beta (Ser9). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
RM453 | |
Unconjugated | |
GSK3B | |
7330414F15Rik; 8430431H08Rik; C86142; EC 2.7.11.26; FA; Factor A; fb68h05; fk80d11; glycogen synthase kinase 3; glycogen synthase kinase 3 beta; glycogen synthase kinase 3 beta L homeolog; glycogen synthase kinase 3 beta variant 1; glycogen synthase kinase 3 beta variant 2; glycogen synthase kinase 3 beta variant 3; glycogen synthase kinase 3 beta variant 4; glycogen synthase kinase 3 beta variant 5; glycogen synthase kinase 3 beta, genome duplicate a; glycogen synthase kinase-3 beta; glycogen synthase kinase-3 beta splice variant X1; glycogen synthase kinase-3 beta splice variant X2; glycogen synthase kinase-3 beta splice variant X3; GSK 3 beta; GSK 3B; gsk3; GSK-3; GSK3 beta; GSK-3 beta; GSK-3 beta GSK3B; GSK-3[b]; Gsk3b; GSK-3b; gsk3b.L; gsk3ba; gsk3beta; GSK-3beta; gsk3-beta; GSK3beta isoform; intracellular kinase; pp37; serine/threonine-protein kinase GSK3B; wu:fb68h05; wu:fk80d11; XELAEV_180112401mg; xgsk-3; Xgsk-3 protein | |
Rabbit | |
Protein A | |
RUO | |
2932 | |
-20°C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.