Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-HAND1 (Ser98) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105803
Description
Antibody detects endogenous levels of HAND1 only when phosphorylated at Ser98.
HAND1 belongs to the basic helix-loop-helix family of transcription factors. This gene product is one of two closely related family members, the HAND proteins, which are asymmetrically expressed in the developing ventricular chambers and play an essential role in cardiac morphogenesis. Working in a complementary fashion, they function in the formation of the right ventricle and aortic arch arteries, implicating them as mediators of congenital heart disease. In addition, it has been suggested that this transcription factor may be required for early trophoblast differentiation.
Specifications
| Phospho-HAND1 (Ser98) | |
| Polyclonal | |
| Unconjugated | |
| HAND1 | |
| bHLHa27; Class A basic helix-loop-helix protein 27; eHAND; Ehand1; extraembryonic tissues, heart, autonomic nervous system and neural crest derivatives-expressed protein 1; HAND1; heart and neural crest derivatives expressed 1; heart and neural crest derivatives expressed transcript 1; heart- and neural crest derivatives-expressed protein 1; Helix-loop-helix transcription factor expressed in extraembryonic mesoderm and trophoblast; Hxt; Th1; Thing1; thing-1 | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 15110, 59112, 9421 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O96004, P97832, Q64279 | |
| HAND1 | |
| A synthesized peptide derived from human HAND1(Accession O96004), corresponding to amino acid residues around phosphorylated Ser98. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction