Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-HBP1 (Ser402) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536809
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536809 Supplier Invitrogen™ Supplier No. PA536809
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 58 and 80 kDa. Purity is >95% by SDS-PAGE.

The HMG-box protein-1 (HBP1) is a member of the HMG family of transcription factors, which are characterized by the presence of a conserved protein motif, the high mobility group (HMG) 1 box, that mediates DNA binding. HBP1 binds to the tumor suppressor proteins Rb and p130 and initiates cell cycle arrest. Terminal cell differentiation requires this initial cell cycle arrest followed by the coordinated expression of genes defined as tissue-specific markers. Along with initiating the commitment to cell differentiation, the continued activity of HBP1 abrogates the expression of tissue-specific genes by associating with the MyoD proteins. In muscle cell differentiation, the MyoD family of transcription factors, which include Myf5, MyoD and myogenein, induce the expression of these cell-type specific proteins and contribute to the development of cell phenotypes. The progression of terminal differentiation is, therefore, dependent on both a decrease in HBP1 activity and the corresponding activation of MyoD-induced gene transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-HBP1 (Ser402)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.2
Gene HBP1
Gene Accession No. O60381, Q62661, Q8R316
Gene Alias 1700058O05Rik; BE629963; C330012F01Rik; C86454; Hbp1; High mobility group box transcription factor 1; high mobility group protein 1; HMG box transcription factor 1; HMG box-containing protein 1; Hmgb1; HMG-box containing protein 1; HMG-box transcription factor 1
Gene Symbols HBP1
Host Species Rabbit
Immunogen Synthetic phosphopeptide derived from human HBP1 around the phosphorylation site of Serine 402.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26959, 27080, 73389
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.