Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599207 Supplier Thermo Scientific Supplier No. PA599207
Add to Cart
Add to Cart

Phospho-HBP1 (Ser402) Polyclonal Antibody

Phospho-HBP1 (Ser402) Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen HBP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HBP1
Gene Accession No. O60381, Q62661, Q8R316
Gene Alias 1700058O05Rik; BE629963; C330012F01Rik; C86454; Hbp1; High mobility group box transcription factor 1; high mobility group protein 1; HMG box transcription factor 1; HMG box-containing protein 1; Hmgb1; HMG-box containing protein 1; HMG-box transcription factor 1
Gene Symbols HBP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HBP1(Accession O60381), corresponding to amino acid residues K355-S405.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26959, 27080, 73389
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.