Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-HCLS1 (Tyr397) Recombinant Rabbit Monoclonal Antibody (HS1Y397-F12), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$245.00 - $616.00
Specifications
Antigen | Phospho-HCLS1 (Tyr397) |
---|---|
Clone | HS1Y397-F12 |
Applications | Flow Cytometry |
Classification | Recombinant Monoclonal |
Conjugate | PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA537407
|
Invitrogen™
MA537407 |
10 Tests |
Each for $245.00
|
|
|||||
PIMA537160
|
Invitrogen™
MA537160 |
100 Tests |
Each for $616.00
|
|
|||||
Description
Substrate of the antigen receptor-coupled tyrosine kinase. Plays a role in antigen receptor signaling for both clonal expansion and deletion in lymphoid cells. May also be involved in the regulation of gene expression. [UniProt]Specifications
Phospho-HCLS1 (Tyr397) | |
Flow Cytometry | |
PE | |
Rabbit | |
Human, Mouse | |
P14317, P49710 | |
15163, 3059 | |
A synthetic phospho-peptide corresponding to residues surrounding Tyr397 of human phospho HS1. | |
Primary | |
4°C | |
HCLS1 |
HS1Y397-F12 | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with 0.20% BSA and 0.09% sodium azide | |
AW213261; CTTNL; HCLS1; hematopoietic cell specific Lyn substrate 1; hematopoietic cell-specific Lyn substrate 1; Hematopoietic lineage cell-specific protein; HS1; LckBP1; p75 | |
HCLS1 | |
IgG κ | |
Protein A/G | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title